Merck
Ledipasvir and sofosbuvir for untreated HCV genotype 1 infection.
The New England journal of medicine (2014-04-15)
Nezam Afdhal, Stefan Zeuzem, Paul Kwo, Mario Chojkier, Norman Gitlin, Massimo Puoti, Manuel Romero-Gomez, Jean-Pierre Zarski, Kosh Agarwal, Peter Buggisch, Graham R Foster, Norbert Bräu, Maria Buti, Ira M Jacobson, G Mani Subramanian, Xiao Ding, Hongmei Mo, Jenny C Yang, Phillip S Pang, William T Symonds, John G McHutchison, Andrew J Muir, Alessandra Mangia, Patrick Marcellin
ABSTRACT
In phase 2 studies, treatment with the all-oral combination of the nucleotide polymerase inhibitor sofosbuvir and the NS5A inhibitor ledipasvir resulted in high rates of sustained virologic response among previously untreated patients with hepatitis C virus (HCV) genotype 1 infection. We conducted a phase 3, open-label study involving previously untreated patients with chronic HCV genotype 1 infection. Patients were randomly assigned in a 1:1:1:1 ratio to receive ledipasvir and sofosbuvir in a fixed-dose combination tablet once daily for 12 weeks, ledipasvir-sofosbuvir plus ribavirin for 12 weeks, ledipasvir-sofosbuvir for 24 weeks, or ledipasvir-sofosbuvir plus ribavirin for 24 weeks. The primary end point was a sustained virologic response at 12 weeks after the end of therapy. Of the 865 patients who underwent randomization and were treated, 16% had cirrhosis, 12% were black, and 67% had HCV genotype 1a infection. The rates of sustained virologic response were 99% (95% confidence interval [CI], 96 to 100) in the group that received 12 weeks of ledipasvir-sofosbuvir; 97% (95% CI, 94 to 99) in the group that received 12 weeks of ledipasvir-sofosbuvir plus ribavirin; 98% (95% CI, 95 to 99) in the group that received 24 weeks of ledipasvir-sofosbuvir; and 99% (95% CI, 97 to 100) in the group that received 24 weeks of ledipasvir-sofosbuvir plus ribavirin. No patient in either 12-week group discontinued ledipasvir-sofosbuvir owing to an adverse event. The most common adverse events were fatigue, headache, insomnia, and nausea. Once-daily ledipasvir-sofosbuvir with or without ribavirin for 12 or 24 weeks was highly effective in previously untreated patients with HCV genotype 1 infection. (Funded by Gilead Sciences; ION-1 ClinicalTrials.gov number NCT01701401.).
Sigma-Aldrich
Uridine 5′-monophosphate, ≥98%
Sigma-Aldrich
Ribavirin, antiviral
Ribavirin, European Pharmacopoeia (EP) Reference Standard