Merck
  • Targeting the androgen receptor with siRNA promotes prostate cancer metastasis through enhanced macrophage recruitment via CCL2/CCR2-induced STAT3 activation.
Targeting the androgen receptor with siRNA promotes prostate cancer metastasis through enhanced macrophage recruitment via CCL2/CCR2-induced STAT3 activation.
EMBO molecular medicine (2013-08-29)
Kouji Izumi, Lei-Ya Fang, Atsushi Mizokami, Mikio Namiki, Lei Li, Wen-Jye Lin, Chawnshang Chang
ABSTRACT
Increased CCL2 expression in prostate cancer (PCa) cells enhanced metastasis via macrophage recruitment. However, its linkage to androgen receptor (AR)-mediated PCa progression remains unclear. Here, we identified a previously unrecognized regulation: targeting AR with siRNA in PCa cells increased macrophage recruitment via CCL2 up-regulation, which might then result in enhancing PCa invasiveness. Molecular mechanism dissection revealed that targeting PCa AR with siRNA promoted PCa cell migration/invasion via CCL2-dependent STAT3 activation and epithelial-mesenchymal transition (EMT) pathways. Importantly, pharmacologic interruption of the CCL2/CCR2-STAT3 axis suppressed EMT and PCa cell migration, providing a new mechanism linking CCL2 and EMT. Simultaneously targeting PCa AR with siRNA and the CCL2/CCR2-STAT3 axis resulted in better suppression of PCa growth and metastasis in a xenograft PCa mouse model. Human PCa tissue microarray analysis suggests that increased CCL2 expression may be potentially associated with poor prognosis of PCa patients. Together, these results may provide a novel therapeutic approach to better battle PCa progression and metastasis at the castration resistant stage via the combination of targeting AR with siRNA and anti-CCL2/CCR2-STAT3 signalling.
Sigma-Aldrich
AG 490, A cell-permeable, reversible, substrate competitive, and potent inhibitor of epidermal growth factor receptor kinase autophosphorylation (IC₅₀ = 100 nM).