Merck
  • A simple biosynthetic method for stereospecific resonance assignment of prochiral methyl groups in proteins.
A simple biosynthetic method for stereospecific resonance assignment of prochiral methyl groups in proteins.
Journal of biomolecular NMR (2011-02-03)
Michael J Plevin, Olivier Hamelin, Jérôme Boisbouvier, Pierre Gans
ABSTRACT
A new method for stereospecific assignment of prochiral methyl groups in proteins is presented in which protein samples are produced using U-[(13)C]glucose and subsaturating amounts of 2-[(13)C]methyl-acetolactate. The resulting non-uniform labeling pattern allows proR and proS methyl groups to be easily distinguished by their different phases in a constant-time two-dimensional (1)H-(13)C correlation spectra. Protein samples are conveniently prepared using the same media composition as the main uniformly-labeled sample and contain higher levels of isotope-enrichment than fractional labeling approaches. This new strategy thus represents an economically-attractive, robust alternative for obtaining isotopically-encoded stereospecific NMR assignments of prochiral methyl groups.
Sigma-Aldrich
(±)-2-Acetoxypropionic acid, ≥97.0% (GC)