Merck
  • Tid1-L inhibits EGFR signaling in lung adenocarcinoma by enhancing EGFR Ubiquitinylation and degradation.
Tid1-L inhibits EGFR signaling in lung adenocarcinoma by enhancing EGFR Ubiquitinylation and degradation.
Cancer research (2013-05-24)
Chi-Yuan Chen, Chia-Ing Jan, Jeng-Fan Lo, Shuenn-Chen Yang, Yih-Leong Chang, Szu-Hua Pan, Wen-Lung Wang, Tse-Ming Hong, Pan-Chyr Yang
ABSTRACT
Tid1 (DNAJA3), a DnaJ cochaperone, may promote degradation of oncogenic kinases. Tid1 has 2 isoforms, Tid1-L and Tid1-S, that may function differently. In this study, we investigated the role of the Tid1 isoforms in regulating EGF receptor (EGFR) signaling and lung cancer progression. We found that both Tid1-L and Tid1-S expressions were reduced in patients with non-small cell lung cancer compared with normal counterparts. Tid1-L expression correlated inversely with EGFR expression. Low Tid1-L/high EGFR expression predicted poor overall survival in patients with lung adenocarcinoma. Tid1-L overexpression in lung cancer cells attenuated EGFR signaling and inhibited cell proliferation, colony formation, and tumor growth in subcutaneous and orthotropic xenograft models. Conversely, depletion of Tid1 restored EGFR signaling and increased cell proliferation and colony formation. Tid1-L, but not Tid1-S, interacted with EGFR/HSP70/HSP90 through the DnaJ domain, counteracting the EGFR regulatory function of HSP90 by causing EGFR ubiquitinylation and proteasomal degradation. Tid1-L inhibited EGFR signaling even more than the HSP90 inhibitor 17-allylamino-demethoxy geldanamycin. We concluded that Tid1-L acted as a tumor suppressor by inhibiting EGFR signaling through interaction with EGFR/HSP70/HSP90 and enhancing EGFR ubiquitinylation and degradation.
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, lyophilized powder, BioReagent, suitable for cell culture