Merck
  • Automated microextraction by packed sorbent of cannabinoids from human urine using a lab-made device packed with molecularly imprinted polymer.
Automated microextraction by packed sorbent of cannabinoids from human urine using a lab-made device packed with molecularly imprinted polymer.
Talanta (2020-09-06)
Douglas Morisue Sartore, Deyber Arley Vargas Medina, Jose Luiz Costa, Fernando Mauro Lanças, Álvaro José Santos-Neto
ABSTRACT
An original, selective and automated method, for the microextraction by packed sorbent (MEPS) of Δ9-tetrahydrocannabinol (THC), 11-hydroxy-Δ9-tetrahydrocannabinol (THC-OH), and 11-nor-Δ9-tetrahydrocannabinol-9-carboxylic acid (THC-COOH) from human urine, was developed by using (i) a catechin-molded molecularly imprinted polymer (MIP), (ii) a new lab-made MEPS device easily repackable with any commercial or lab-made sorbent, and (iii) a lab-made multi syringe autosampler. Analyses were performed by liquid chromatography-tandem mass spectrometry (LC-MS/MS) and the developed method proved to be precise, accurate and showed good linearity. Determination coefficients ranged from 0.96 to 0.99, in the range of 5-250 ng mL-1. Limits of detection and quantification ranged between 1.0 and 5.0 ng mL-1 and 5.0 and 20.0 ng mL-1. The method was successfully applied in the analysis of real urine samples. The same packed syringe was effectively used over 90 consecutive extractions without carry-over effects.
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene
Sigma-Aldrich
2,2′-Azobis(2-methylpropionamidine) dihydrochloride, powder or granules, 97%