Merck
  • Cellular toxicological characterization of a thioxolated arsenic-containing hydrocarbon.
Cellular toxicological characterization of a thioxolated arsenic-containing hydrocarbon.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS) (2020-06-13)
Franziska Ebert, Vanessa Ziemann, Viktoria Klara Wandt, Barbara Witt, Sandra Marie Müller, Nikolaus Guttenberger, Ezgi Eyluel Bankoglu, Helga Stopper, Georg Raber, Kevin A Francesconi, Tanja Schwerdtle
ABSTRACT
Arsenolipids, especially arsenic-containing hydrocarbons (AsHC), are an emerging class of seafood originating contaminants. Here we toxicologically characterize a recently identified oxo-AsHC 332 metabolite, thioxo-AsHC 348 in cultured human liver (HepG2) cells. Compared to results of previous studies of the parent compound oxo-AsHC 332, thioxo-AsHC 348 substantially affected cell viability in the same concentration range but exerted about 10-fold lower cellular bioavailability. Similar to oxo-AsHC 332, thioxo-AsHC 348 did not substantially induce oxidative stress nor DNA damage. Moreover, in contrast to oxo-AsHC 332 mitochondria seem not to be a primary subcellular toxicity target for thioxo-AsHC 348. This study indicates that thioxo-AsHC 348 is at least as toxic as its parent compound oxo-AsHC 332 but very likely acts via a different mode of toxic action, which still needs to be identified.
Sigma-Aldrich
7-Amino-4-(trifluoromethyl)coumarin, ≥99%