
Ubidecarenone

Synonym(s):
Coenzyme Q10, Q-10, Ubiquinone 50, Ubiquinone-10
Empirical Formula (Hill Notation):
C59H90O4
CAS Number:
Molecular Weight:
863.34
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique