
Promazine hydrochloride

Synonym(s):
10-(3-Dimethylaminopropyl)phenothiazine hydrochloride, Promazine hydrochloride
Linear Formula:
C17H20N2S · HCl
CAS Number:
Molecular Weight:
320.88
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing