
Primidone Related Compound C

Synonym(s):
2-Phenylbutyramide, α-Ethylbenzeneacetamide
Empirical Formula (Hill Notation):
C10H13NO
CAS Number:
Molecular Weight:
163.22
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2