
Pentamidine isethionate

Synonym(s):
Pentamidine isethionate salt, 1,5-Bis(p-amidinophenoxy)pentane bis(2-hydroxyethanesulfonate salt), 4,4′-(Pentamethylenedioxy)dibenzamidine bis(2-hydroxyethanesulfonate)
Linear Formula:
CH2[CH2CH2OC6H4C(=NH)NH2]2·2HOCH2CH2SO3H
CAS Number:
Molecular Weight:
592.68
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing