
Nomegestrol acetate for ID and assay CRS

Synonym(s):
Nomegestrol acetate, 17-Acetoxy-6-methyl-19-norpregna-4,6-diene-3,20-dione, NOMAc, Org 10486-0
Empirical Formula (Hill Notation):
C23H30O4
CAS Number:
Molecular Weight:
370.48
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing