Skip to Content
Merck
Galangin phyproof® Reference Substance

Galangin

Galangin phyproof® Reference Substance
Synonym(s):
3,5,7-Trihydroxyflavone, Norizalpinin
Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
EC Number:

Products

Product Category
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing