
Clethodim

Synonym(s):
2-{(E)-1-[(E)-3-Chloroallyloxyimino]propyl}-5-[2-(ethylthio)propyl]-3-hydroxy-2-cyclohexen-1-one
Empirical Formula (Hill Notation):
C17H26ClNO3S
CAS Number:
Molecular Weight:
359.91
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2