
Cefazolin Impurity E

Synonym(s):
5-Methyl-1,3,4-thiadiazole-2-thiol, 2-Mercapto-5-methyl-1,3,4-thiadiazole
Empirical Formula (Hill Notation):
C3H4N2S2
CAS Number:
Molecular Weight:
132.21
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing