HPLC Analysis of Carbamate and Urea Pesticides on SUPELCOSIL™ LC-8
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SUPELCOSIL LC-8, 15 cm × 4.6 mm I.D., 5 μm particles (58220-U)
mobile phase
[A] acetonitrile; [B] water
gradient
18:82 to 65:35 in 9 min, hold 3 min
flow rate
2 mL/min
column temp.
35 °C
detector
UV, 240 nm
injection
10 μL
Description
Legal Information
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC