US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
CONDITIONS
column
SLB-5ms, 20 m x 0.18 mm I.D., 0.36 μm (28576-U)
oven
40 °C (1 min), 12 °C/min to 200 °C, 20 °C/min to 340 °C (2 min)
inj. temp.
250 °C
MSD interface
340 °C
scan range
m/z 40-450
carrier gas
helium, programmed flow; 0.5 mL/min (5 min hold), to 1 mL/min at 5.5 min (constant for remainder of run)
injection
0.50 μl, pulsed splitless (25 psi until 0.1 min) (0.75 min)
liner
4 mm I.D., single taper
sample
80 component semivolatile standard at 50 ppm plus 6 internal standards (at 40 ppm) in methylene chloride
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany