GC Analysis of Volatiles in Water on SPB®-1 after SPME using 100 μm PDMS Fiber
CONDITIONS
SPME fiber
polydimethylsiloxane, 100 μm (57300-U)
extraction
5 min immersed in water with rapid stirring
column
SPB-1, 10 m × 0.20 mm I.D., 1.2 μm (24134-U)
oven
40 °C ( 0.75 min) to 160 °C at 20 °C/min
inj. temp.
230 °C, splitless (closed 3 min)
detector
FID, 260 °C
carrier gas
helium, 40 cm/sec
sample
50ppb each analyte in 1.8 mL saturated salt water
Description
Legal Information
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany