Merck
GC Analysis of Volatiles in Multifloral Honey on SUPELCOWAX 10 after SPME using a 50/30 μm DVB/Carboxen/PDMS Fiber
GC Analysis of Volatiles in Multifloral Honey on SUPELCOWAX 10 after SPME using a 50/30 μm DVB/Carboxen/PDMS Fiber suitable for GC, application for SPME
Materials
analytical column
Product No.
Description
Pricing
SUPELCOWAX 10 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SPME fiber
Product No.
Description
Pricing
SPME Fiber Assembly
50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
related product
Product No.
Description
Pricing
SPME Fiber Holder
for use with Manual Sampling
SPME Fiber Holder
for use with CTC CombiPAL, Gerstel MPS2
standard
Product No.
Description
Pricing
Decanal
≥98% (GC), liquid
Nonanoic acid
analytical standard
Benzaldehyde
analytical standard
Phenylacetaldehyde
≥90%
Polar Column Test Mix
certified reference material, 500 μg/mL each component in dichloromethane
Nonanal
analytical standard
Phenylethyl Alcohol
Pharmaceutical Secondary Standard; Certified Reference Material
Furfural
analytical standard
CONDITIONS
sample/matrix
5 g multifloral honey + 5 mL bidistilled water in a 20 mL vial, allowed to equilibrate at 50 °C for 15 min with magnetic stirring
SPME fiber
2 cm 50/30 μm divinylbenzene/carboxen on polydimethylsiloxane (57348-U)
extraction
headspace, 50 °C for 40 min with magnetic stirring
desorption process
250 °C for 2 min
column
SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 μm (24079)
oven
35 °C (8 min), 6 °C/min to 60 °C, 4 °C/min to 160 °C, 20 °C/min to 200 °C (1 min)
inj. temp.
250 °C
MSD interface
230 °C
scan range
35-300 m/z
carrier gas
helium, 1 mL/min
liner
0.75 mm I.D., SPME
sample
(Honey)
Description
Analysis Note
In this application, headspace SPME (HS-SPME) using a Carboxen/PDMS/DVB fiber coupled with gas chromatography-mass spectrometry (GC/MS) was used for the characterization of the volatile fraction of honey samples. Using SPME it was possible to obtain and distinguish the chemical fingerprints of different types of honey. The relatively polar SUPELCOWAX 10 GC column provides excellent separation of these polar compounds.
Legal Information
null