Merck
  • Bimane fluorescent labels: labeling of normal human red cells under physiological conditions.
Bimane fluorescent labels: labeling of normal human red cells under physiological conditions.
Proceedings of the National Academy of Sciences of the United States of America (1979-07-01)
N S Kosower, E M Kosower, G L Newton, H M Ranney
PMID291011
ABSTRACT
The bimane fluorescent labels, monobromobimane, dibromobimane, and monobromotrimethylammoniobimane, are derivatives of syn-9,10-dioxabimane:1,5-diazabicyclo[3.3.0]octa-3,6-diene-2,8-dione. They efficiently label hemoglobin (reactive thiol groups), membrane proteins, and glutathione of normal human red cells under physiological conditions. Monobromobimane and dibromobimane are effective on intact cells while red cell membranes may be impermeable to the positively charged monobromotrimethylammoniobimane, the latter being effective only on lysed cells. These bimane labels provide a class of labeling agents that may have wide applicability in biological materials.
Sigma-Aldrich
Bromobimane, ≥97% (HPLC)
Pricing and availability is not currently available.