- The mitochondrial Na
The mitochondrial Na
Nature (2017-04-27)
Timothy S Luongo, Jonathan P Lambert, Polina Gross, Mary Nwokedi, Alyssa A Lombardi, Santhanam Shanmughapriya, April C Carpenter, Devin Kolmetzky, Erhe Gao, Jop H van Berlo, Emily J Tsai, Jeffery D Molkentin, Xiongwen Chen, Muniswamy Madesh, Steven R Houser, John W Elrod
PMID28445457
ABSTRACT
Mitochondrial calcium (
Sigma-Aldrich
Anti-MCU antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution