Merck
  • Novel MYBL1 Gene Rearrangements with Recurrent MYBL1-NFIB Fusions in Salivary Adenoid Cystic Carcinomas Lacking t(6;9) Translocations.
Novel MYBL1 Gene Rearrangements with Recurrent MYBL1-NFIB Fusions in Salivary Adenoid Cystic Carcinomas Lacking t(6;9) Translocations.
Clinical cancer research : an official journal of the American Association for Cancer Research (2015-12-04)
Yoshitsugu Mitani, Bin Liu, Pulivarthi H Rao, Vishnupriya J Borra, Mark Zafereo, Randal S Weber, Merrill Kies, Guillermina Lozano, P Andrew Futreal, Carlos Caulin, Adel K El-Naggar
ABSTRACT
Adenoid cystic carcinoma (ACC) is an indolent salivary gland malignancy, characterized by t(6;9) translocations and MYB-NFIB gene fusions in approximately 50% of the tumors. The genetic alterations underlying t(6;9)-negative and t(6;9)-positive/MYB-NFIB fusion-negative ACC remain unknown. To uncover the genetic alterations in ACC lacking the canonical translocation and fusion transcript and identify new abnormalities in translocation positive tumors. We performed whole-genome sequencing in 21 salivary ACCs and conducted targeted molecular analyses in a validation set (81 patients). Microarray gene-expression data were also analyzed to explore the biologic differences between fusion positive and negative tumors. We identified a novel MYBL1-NFIB gene fusion as a result of t(8;9) translocation and multiple rearrangements in the MYBL1 gene in 35% of the t(6;9)-negative ACCs. All MYBL1 alterations involved deletion of the C-terminal negative regulatory domain and were associated with high MYBL1 expression. Reciprocal MYB and MYBL1 expression was consistently found in ACCs. In addition, 5'-NFIB fusions that did not involve MYB/MYBL1 genes were identified in a subset of t(6;9)-positive/fusion-negative tumors. We also delineated distinct gene-expression profiles in ACCs associated with the length of the MYB or MYBL1 fusions, suggesting a biologic importance of the C-terminal part of these fusions. Our study defines new molecular subclasses of ACC characterized by MYBL1 rearrangements and 5'-NFIB gene fusions.