Merck
  • STA-21, a STAT-3 inhibitor, attenuates the development and progression of inflammation in collagen antibody-induced arthritis.
STA-21, a STAT-3 inhibitor, attenuates the development and progression of inflammation in collagen antibody-induced arthritis.
Immunobiology (2016-10-09)
Sheikh Fayaz Ahmad, Mushtaq Ahmad Ansari, Ahmed Nadeem, Khairy M A Zoheir, Saleh A Bakheet, Abdulaziz M S Alsaad, Othman A Al-Shabanah, Sabry M Attia
ABSTRACT
We set out to investigate the influence of STA-21, a dynamic STAT-3 inhibitor, on the expansion and progression of rheumatoid arthritis (RA), and to determine its potential mechanisms of action in a mouse model of collagen antibody-induced arthritis (CAIA). To this end, arthritis was induced via intravenous (IV) injection of Balb/c mice with a cocktail of antibodies directed against type II collagen (1.5μg/mouse, IV), followed by lipopolysaccharide (LPS) at a dose of (25μg/mouse, i.p.) on day 3. Mice were then left untreated or were simultaneously treated with STA-21 (0.5mg/kg, i.p., once daily for 2 weeks) followed by evaluation for clinical and histological features of arthritic inflammation and flow cytometric analysis of cytokines and transcription factors in peripheral blood. STA-21 enhanced the clinical course of arthritis in CAIA mice and decreased CD8