Merck
  • Therapeutic potential and mechanism of kinetin as a treatment for the human splicing disease familial dysautonomia.
Therapeutic potential and mechanism of kinetin as a treatment for the human splicing disease familial dysautonomia.
Journal of molecular medicine (Berlin, Germany) (2007-01-09)
Matthew M Hims, El Chérif Ibrahim, Maire Leyne, James Mull, Lijuan Liu, Conxi Lazaro, Ranjit S Shetty, Sandra Gill, James F Gusella, Robin Reed, Susan A Slaugenhaupt
ABSTRACT
Mutations that affect the splicing of pre-mRNA are a major cause of human disease. Familial dysautonomia (FD) is a recessive neurodegenerative disease caused by a T to C transition at base pair 6 of IKBKAP intron 20. This mutation results in variable tissue-specific skipping of exon 20. Previously, we reported that the plant cytokinin kinetin dramatically increases exon 20 inclusion in RNA isolated from cultured FD cells. The goal of the current study was to investigate the nature of the FD splicing defect and the mechanism by which kinetin improves exon inclusion, as such knowledge will facilitate the development of future therapeutics aimed at regulating mRNA splicing. In this study, we demonstrate that treatment of FD lymphoblast cell lines with kinetin increases IKBKAP mRNA and IKAP protein to normal levels. Using a series of minigene constructs, we show that deletion of a region at the end of IKBKAP exon 20 disrupts the ability of kinetin to improve exon inclusion, pinpointing a kinetin responsive sequence element. We next performed a screen of endogenously expressed genes with multiple isoforms resulting from exon skipping events and show that kinetin's ability to improve exon inclusion is not limited to IKBKAP. Lastly, we highlight the potential of kinetin for the treatment of other human splicing disorders by showing correction of a splicing defect in neurofibromatosis.
Sigma-Aldrich
Kinetin, ≥99.0% (HPLC)
Pricing and availability is not currently available.