Merck
  • Percutaneous cholecystostomy in the management of high-risk patients presenting with acute cholecystitis: Timing and outcome at a single institution.
Percutaneous cholecystostomy in the management of high-risk patients presenting with acute cholecystitis: Timing and outcome at a single institution.
American journal of surgery (2017-02-27)
Shiri Zarour, Ashraf Imam, Guennadi Kouniavsky, Guy Lin, Andrew Zbar, Eli Mavor
ABSTRACT
Cholecystectomy is the standard of care in acute cholecystitis (AC). Percutaneous cholecystostomy (PC) is an effective alternative for high-risk surgical cases. A retrospective analysis is presented of AC patients treated with PC drainage at a single tertiary institution over a 21 month period, assessing outcome and complications. Of 119 patients, 103 had clinical improvement after PC insertion. There were 7 peri-procedural deaths (5.9%), all in elderly high-risk cases. Overall, 56/103 cases (54%) were definitively managed with PC drainage with 41 patients (40%) undergoing an elective cholecystectomy (75% performed laparoscopically). The timing of PC insertion did not affect AC resolution or drain-related complications, although more patients underwent an elective cholecystectomy if PC placement was delayed (>24 h after admission). In AC, drainage by a PC catheter is a safe and effective procedure. It may be used either as a bridge to elective cholecystectomy or in selected cases as definitive therapy.
Product Description
Supelco
Cefuroxime, VETRANAL®, analytical standard
Pricing and availability is not currently available.