Merck
  • Enhancement of the activity of phenoxodiol by cisplatin in prostate cancer cells.
Enhancement of the activity of phenoxodiol by cisplatin in prostate cancer cells.
British journal of cancer (2009-02-12)
R A C McPherson, P T Galettis, P L de Souza
ABSTRACT
Phenoxodiol is a novel isoflav-3-ene, currently undergoing clinical trials, that has a broad in vitro activity against a number of human cancer cell lines. Phenoxodiol alone inhibited DU145 and PC3 in a dose- and time-dependent manner with IC(50) values of 8+/-1 and 38+/-9 microM, respectively. The combination of phenoxodiol and cisplatin was synergistic in DU145, and additive in PC3, as assessed by the Chou-Talalay method. Carboplatin was also synergistic in combination with phenoxodiol in DU145 cells. The activity of the phenoxodiol and cisplatin combination was confirmed in vivo using a DU145 xenograft model in nude mice. Pharmacokinetic data from these mice suggest that the mechanism of synergy may occur through a pharmacodynamic mechanism. An intracellular cisplatin accumulation assay showed a 35% (P<0.05) increase in the uptake of cisplatin when it was combined in a ratio of 1 microM:5 microM phenoxodiol, resulting in a 300% (P<0.05) increase in DNA adducts. Taken together, our results suggest that phenoxodiol has interesting properties that make combination therapy with cisplatin or carboplatin appealing.
Sigma-Aldrich
Phenoxodiol, ≥98% (HPLC)