Merck
  • Arrested and temporarily arrested states in a protein-polymer mixture studied by USAXS and VSANS.
Arrested and temporarily arrested states in a protein-polymer mixture studied by USAXS and VSANS.
Soft matter (2017-11-14)
Stefano Da Vela, Christian Exner, Richard Santiago Schäufele, Johannes Möller, Zhendong Fu, Fajun Zhang, Frank Schreiber
ABSTRACT
We investigate the transition of the phase separation kinetics from a complete to an arrested liquid-liquid phase separation (LLPS) in mixtures of bovine γ-globulin with polyethylene glycol (PEG). The solutions feature LLPS with upper critical solution temperature phase behavior. At higher PEG concentrations or low temperatures, non-equilibrium, gel-like states are found. The kinetics is followed during off-critical quenches by ultra-small angle X-ray scattering (USAXS) and very-small angle neutron scattering (VSANS). For shallow quenches a kinetics consistent with classical spinodal decomposition is found, with the characteristic length (ξ) growing with time as ξ ∼ t