Merck
  • SuFEx-Based Polysulfonate Formation from Ethenesulfonyl Fluoride-Amine Adducts.
SuFEx-Based Polysulfonate Formation from Ethenesulfonyl Fluoride-Amine Adducts.
Angewandte Chemie (International ed. in English) (2017-08-10)
Hua Wang, Feng Zhou, Gerui Ren, Qinheng Zheng, Hongli Chen, Bing Gao, Liana Klivansky, Yi Liu, Bin Wu, Qingfeng Xu, Jianmei Lu, K Barry Sharpless, Peng Wu
ABSTRACT
The SuFEx-based polycondensation between bisalkylsulfonyl fluorides (AA monomers) and bisphenol bis(t-butyldimethylsilyl) ethers (BB monomers) using [Ph
Sigma-Aldrich
Tris(dimethylamino)sulfonium bifluoride
Sigma-Aldrich
Bis(triphenyl-λ5-phosphanylidene)ammonium hydrogendifluoride