Merck
  • Structurally Analogous Degradable Version of Fluorene-Bipyridine Copolymer with Exceptional Selectivity for Large-Diameter Semiconducting Carbon Nanotubes.
Structurally Analogous Degradable Version of Fluorene-Bipyridine Copolymer with Exceptional Selectivity for Large-Diameter Semiconducting Carbon Nanotubes.
ACS applied materials & interfaces (2017-10-27)
Catherine Kanimozhi, Gerald J Brady, Matthew J Shea, Peishen Huang, Yongho Joo, Michael S Arnold, Padma Gopalan
ABSTRACT
Separation of electronically pure, narrowly dispersed, pristine, semiconducting single-walled carbon nanotubes (CNTs) from a heterogeneous as-synthesized mixture is essential for various semiconducting technologies and biomedical applications. Although conjugated polymer wrappers are often utilized to facilitate electronic-type sorting, it is highly desirable to remove organic residues from the resulting devices. We report here the design and synthesis of a mild acid-degradable π-conjugated polyimine polymer, poly[(9,9-di-n-octyl-2,7-fluoren-dinitrilomethine)-alt-co-(6,6'-{2,2'-bipyridyl-dimethine})] (PFO-N-BPy), that is structurally analogous to the commonly used and commercially available poly[(9,9-dioctylfluorenyl-2,7-diyl)-alt-co-(6,6'-(2,2'-bipyridine))] (PFO-BPy). An acid cleavable imine link (-HC═N-) was introduced in the PFO-N-BPy backbone to impart degradability, which is absent in PFO-BPy. PFO-N-BPy was synthesized via a metal catalyst-free aza-Wittig reaction in high yields. PFO-N-BPy with a degree of polymerization of just ∼10 showed excellent (>99% electronic purity) selectivity for both large-diameter (1.3-1.7 nm) arc-discharge semiconducting CNTs (S-CNTs) and smaller diameter (0.8-1.2 nm) high-pressure carbon monoxide disproportionation reaction S-CNTs. Overall, the selectivity for the semiconducting species is similar to that of PFO-BPy but with an advantage of complete depolymerization under mild acidic conditions into recyclable monomers. We further show by ultraviolet-visible spectroscopy, X-ray photoelectron spectroscopy, and scanning electron microscopy that the PFO-N-BPy-wrapped S-CNTs can be aligned into a monolayer array on gate dielectrics using a floating evaporative self-assembly process from which the polymer can be completely removed. Short channel field effect transistors were fabricated from the polymer-stripped aligned S-CNT arrays, which further confirmed the semiconducting purity on the order of 99.9% or higher.
Sigma-Aldrich
Carbon nanotube, single-walled, >70 wt. % carbon basis, D × L 2-10 nm × 1-5 μm , bundle dimensions, avg. no. of layers, 1
Pricing and availability is not currently available.