Merck
Dual-pH/Magnetic-Field-Controlled Drug Delivery Systems Based on Fe
ChemMedChem (2017-09-01)
Xinyu Hu, Yongmei Wang, Liangliang Zhang, Man Xu, Jianfa Zhang, Wei Dong
ABSTRACT
Salecan is a water-soluble extracellular β-glucan and has excellent physicochemical and biological properties for hydrogel preparation. In this study, a new pH/magnetic field dual-responsive hydrogel was prepared by the graft copolymerization of salecan with 4-pentenoic acid (PA) and N-hydroxyethylacrylamide (HEAA) in the presence of Fe
Sigma-Aldrich
N-Hydroxyethyl acrylamide, contains 1,000 ppm monomethyl ether hydroquinone as stabilizer, 97%
Pricing and availability is not currently available.