Merck
  • AQP4-knockout aggravation of isoprenaline-induced myocardial injury is mediated by p66Shc and endoplasmic reticulum stress.
AQP4-knockout aggravation of isoprenaline-induced myocardial injury is mediated by p66Shc and endoplasmic reticulum stress.
Clinical and experimental pharmacology & physiology (2017-07-09)
Yusi Cheng, Jie Chao, Dezai Dai, Yin Dai, Dongdong Zhu, Bicheng Liu
ABSTRACT
Aquaporin 4 (AQP4) is a type of water channel protein that maintains the water balance of cardiomyocytes. However, the physiological role of AQP4 in cardiovascular disease is poorly understood. We wanted to explore whether p66Shc and endoplasmic reticulum stress participates in AQP4 knockout (KO)-mediated cardiac injury. There were two types of mice: AQP4 knockout and wild-type mice. Each type was randomly divided into three groups: Control group, isoprenaline stimulation group (ISO, 1 mg/kg, s.c., 5 days), and apocynin treatment group (APO, 100 mg/kg, p.o., 3 days). H9c2 rat cardiomyocytes were cultured for RNA interference of AQP4. Results showed increased left ventricular weight index and more severe myocardial inflammation were induced in AQP4 knockout mice relative to wild-type mice, accompanied by significantly increased levels of the oxidative stress biomarkers MDA and NOX4. In addition, the expressions of p66Shc, ER stress markers PERK, GRP78 and CHOP and proinflammatory factors such as ET