Merck
  • Tubuloglomerular feedback responses in offspring of dexamethasone-treated ewes.
Tubuloglomerular feedback responses in offspring of dexamethasone-treated ewes.
American journal of physiology. Renal physiology (2017-07-07)
Anita J Turner, Russell D Brown, Amanda E Brandon, A Erik G Persson, Karen J Gibson
ABSTRACT
Via developmental programming, prenatal perturbations, such as exposure to glucocorticoids and maternal malnutrition alter kidney development and contribute to the development of hypertension. To examine the possibility that alterations in tubuloglomerular feedback (TGF) contribute to the development of hypertension in offspring following maternal dexamethasone treatment (Dex) in early gestation, studies were conducted in fetal sheep and lambs. Pregnant ewes were infused with dexamethasone (0.48 mg/h) at 26-28 days gestation. No differences were observed in mean arterial pressure, glomerular filtration rate. or electrolyte excretion rates between the Dex and Untreated fetuses or lambs. Gestational exposure to Dex markedly enhanced TGF sensitivity, as the turning point in Dex-treated fetuses was significantly lower (12.9 ± 0.9 nl/min;
Sigma-Aldrich
7-Nitroindazole, ≥98%
Pricing and availability is not currently available.