Merck
Kinetic properties of the mitochondrial F
Biochimie (2017-06-28)
Salvatore Nesci, Fabiana Trombetti, Vittoria Ventrella, Maurizio Pirini, Alessandra Pagliarani
ABSTRACT
The mitochondrial F-ATPase can be activated either by the classical cofactor Mg
Sigma-Aldrich
2,5-Di-tert-butylhydroquinone, 99%
Pricing and availability is not currently available.