Merck
C-H Xanthylation: A Synthetic Platform for Alkane Functionalization.
Journal of the American Chemical Society (2016-10-16)
William L Czaplyski, Christina G Na, Erik J Alexanian
ABSTRACT
Intermolecular functionalizations of aliphatic C-H bonds offer unique strategies for the synthesis and late-stage derivatization of complex molecules, but the chemical space accessible remains limited. Herein, we report a transformation significantly expanding the chemotypes accessible via C-H functionalization. The C-H xanthylation proceeds in useful chemical yields with the substrate as the limiting reagent using blue LEDs and an easily prepared N-xanthylamide. The late-stage functionalizations of complex molecules occur with high levels of site selectivity, and a variety of common functionality is tolerated in the reaction. This approach capitalizes on the versatility of the xanthate functional group via both polar and radical manifolds to unlock a wide array of C-H transformations previously inaccessible in synthesis.
Sigma-Aldrich
N-(tert-Butyl)-N-((ethoxycarbonothioyl)thio)-3,5-bis(trifluoromethyl)benzamide
Pricing and availability is not currently available.