Merck
Tracking the insulator-to-metal phase transition in VO
Proceedings of the National Academy of Sciences of the United States of America (2017-08-23)
Marieke F Jager, Christian Ott, Peter M Kraus, Christopher J Kaplan, Winston Pouse, Robert E Marvel, Richard F Haglund, Daniel M Neumark, Stephen R Leone
ABSTRACT
Coulomb correlations can manifest in exotic properties in solids, but how these properties can be accessed and ultimately manipulated in real time is not well understood. The insulator-to-metal phase transition in vanadium dioxide (VO
Sigma-Aldrich
Vanadium(IV) oxide, ≥99% trace metals basis
Pricing and availability is not currently available.