Merck
Activation of the CREB/
Frontiers in cellular neuroscience (2017-07-14)
Daniela Gandolfi, Silvia Cerri, Jonathan Mapelli, Mariarosa Polimeni, Simona Tritto, Marie-Therese Fuzzati-Armentero, Albertino Bigiani, Fabio Blandini, Lisa Mapelli, Egidio D'Angelo
ABSTRACT
The induction of long-term potentiation and depression (LTP and LTD) is thought to trigger gene expression and protein synthesis, leading to consolidation of synaptic and neuronal changes. However, while LTP and LTD have been proposed to play important roles for sensori-motor learning in the cerebellum granular layer, their association with these mechanisms remained unclear. Here, we have investigated phosphorylation of the cAMP-responsive element binding protein (CREB) and activation of the immediate early gene
Sigma-Aldrich
4-(2-(6-(Dibutylamino)-2-naphthalenyl)ethenyl)-1-(3-sulfopropyl)pyridinium hydroxide inner salt, ≥95% (HPLC), solid
Sigma-Aldrich
D(−)-2-Amino-5-phosphonopentanoic acid, NMDA receptor antagonist