Merck
Analytical approaches to investigate protein-pesticide adducts.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2009-11-03)
Wayne G Carter, Mabruka H Tarhoni, David E Ray
ABSTRACT
Organophosphorus pesticides primarily elicit toxicity via their common covalent adduction of acetylcholinesterase (AChE), but pesticide binding to additional sensitive secondary targets may also compromise health. We have utilised tritiated-diisopropylfluorophosphate ((3)H-DFP) binding to quantify the levels of active immune and brain tissue serine hydrolases, and visualise them using autoradiography after protein separation by one-dimensional and two-dimensional techniques. Preincubation of protein extracts with pesticide in vitro or dosing of rats with pesticide in vivo was followed by (3)H-DFP radiolabelling. Pesticide targets were identified by a reduction in (3)H-DFP radiolabelling relative to controls, and characterised by their tissue presence, molecular weight, and isoelectric point. Conventional column chromatography was employed to enrich pesticide targets to enable their further characterisation, and/or identification by mass spectrometry. The major in vivo pesticide targets characterised were 66 kDa, serum albumin, and 60 kDa, likely carboxylesterase 1, both of which displayed differential pesticide binding character under conditions producing approximately 30% tissue AChE inhibition. The characterisation and identification of sensitive pesticide secondary targets will enable an evaluation of their potential contribution to the ill health that may arise from chronic low-dose pesticide exposures. Additionally, secondary targets may provide useful biomonitors and/or bioscavengers of pesticide exposures.
Sigma-Aldrich
Acetylcholinesterase human, recombinant, expressed in HEK 293 cells, lyophilized powder, ≥1,000 units/mg protein (Lowry)