Merck
  • Glaucoma: Biological Trabecular and Neuroretinal Pathology with Perspectives of Therapy Innovation and Preventive Diagnosis.
Glaucoma: Biological Trabecular and Neuroretinal Pathology with Perspectives of Therapy Innovation and Preventive Diagnosis.
Frontiers in neuroscience (2017-09-21)
Raffaele Nuzzi, Federico Tridico
ABSTRACT
Glaucoma is a common degenerative disease affecting
Sigma-Aldrich
R(+)-Methanandamide, 5 mg/mL in absolute ethanol, ≥96% (HPLC)