- Pharmacological inhibition of CaMKK2 with the selective antagonist STO-609 regresses NAFLD.
Pharmacological inhibition of CaMKK2 with the selective antagonist STO-609 regresses NAFLD.
Scientific reports (2017-09-20)
Brian York, Feng Li, Fumin Lin, Kathrina L Marcelo, Jianqiang Mao, Adam Dean, Naomi Gonzales, David Gooden, Suman Maity, Cristian Coarfa, Nagireddy Putluri, Anthony R Means
PMID28924233
ABSTRACT
Binding of calcium to its intracellular receptor calmodulin (CaM) activates a family of Ca
Sigma-Aldrich
CAMKK2, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution