- Novel MYC-driven medulloblastoma models from multiple embryonic cerebellar cells.
Novel MYC-driven medulloblastoma models from multiple embryonic cerebellar cells.
Oncogene (2017-05-16)
D Kawauchi, R J Ogg, L Liu, D J H Shih, D Finkelstein, B L Murphy, J E Rehg, A Korshunov, C Calabrese, F Zindy, T Phoenix, Y Kawaguchi, J Gronych, R J Gilbertson, P Lichter, A Gajjar, M Kool, P A Northcott, S M Pfister, M F Roussel
PMID28504719
ABSTRACT
Group3 medulloblastoma (MB