Merck
  • Merging photoredox and nickel catalysis: decarboxylative cross-coupling of carboxylic acids with vinyl halides.
Merging photoredox and nickel catalysis: decarboxylative cross-coupling of carboxylic acids with vinyl halides.
Journal of the American Chemical Society (2014-12-19)
Adam Noble, Stefan J McCarver, David W C MacMillan
ABSTRACT
Decarboxylative cross-coupling of alkyl carboxylic acids with vinyl halides has been accomplished through the synergistic merger of photoredox and nickel catalysis. This new methodology has been successfully applied to a variety of α-oxy and α-amino acids, as well as simple hydrocarbon-substituted acids. Diverse vinyl iodides and bromides give rise to vinylation products in high efficiency under mild, operationally simple reaction conditions.
Sigma-Aldrich
[Ir(dF(Me)ppy)2(dtbbpy)]PF6
Pricing and availability is not currently available.