Merck
  • Neurophysiology of space travel: energetic solar particles cause cell type-specific plasticity of neurotransmission.
Neurophysiology of space travel: energetic solar particles cause cell type-specific plasticity of neurotransmission.
Brain structure & function (2016-12-03)
Sang-Hun Lee, Barna Dudok, Vipan K Parihar, Kwang-Mook Jung, Miklós Zöldi, Young-Jin Kang, Mattia Maroso, Allyson L Alexander, Gregory A Nelson, Daniele Piomelli, István Katona, Charles L Limoli, Ivan Soltesz
ABSTRACT
In the not too distant future, humankind will embark on one of its greatest adventures, the travel to distant planets. However, deep space travel is associated with an inevitable exposure to radiation fields. Space-relevant doses of protons elicit persistent disruptions in cognition and neuronal structure. However, whether space-relevant irradiation alters neurotransmission is unknown. Within the hippocampus, a brain region crucial for cognition, perisomatic inhibitory control of pyramidal cells (PCs) is supplied by two distinct cell types, the cannabinoid type 1 receptor (CB
Sigma-Aldrich
Glycerol-d8, ≥98 atom % D, ≥98% (CP)
Pricing and availability is not currently available.