- Electrocatalytic dechlorination of 2,3,5-trichlorophenol on palladium/carbon nanotubes-nafion film/titanium mesh electrode.
Electrocatalytic dechlorination of 2,3,5-trichlorophenol on palladium/carbon nanotubes-nafion film/titanium mesh electrode.
Environmental science and pollution research international (2017-04-22)
Zhirong Sun, Xiaoyue Ma, Xiang Hu
PMID28429272
ABSTRACT
Palladium/carbon nanotubes-nafion film-modified titanium mesh electrode (Pd/CNTs-nafion film/Ti electrode) was prepared and used for catalytic dechlorination of 2,3,5-trichlorophenol (2,3,5-TCP). The influences of factors, such as Pd
MATERIALS