Merck
  • CXCL17 Attenuates Imiquimod-Induced Psoriasis-like Skin Inflammation by Recruiting Myeloid-Derived Suppressor Cells and Regulatory T Cells.
CXCL17 Attenuates Imiquimod-Induced Psoriasis-like Skin Inflammation by Recruiting Myeloid-Derived Suppressor Cells and Regulatory T Cells.
Journal of immunology (Baltimore, Md. : 1950) (2017-04-09)
Tomonori Oka, Makoto Sugaya, Naomi Takahashi, Takehiro Takahashi, Sayaka Shibata, Tomomitsu Miyagaki, Yoshihide Asano, Shinichi Sato
ABSTRACT
CXCL17 is expressed in a variety of cancers and promotes tumor progression by recruiting myeloid-derived suppressor cells (MDSCs). MDSCs suppress tumor immunity by attracting regulatory T cells (Tregs) into tumor sites through CCL5. In this study, we examined the role of CXCL17 in skin disorders. CXCL17 mRNA levels in psoriasis skin, but not in lesional skin of atopic dermatitis or cutaneous T cell lymphoma, were significantly higher than those in normal skin. CXCL17 was mainly expressed in the epidermis, and IFN-γ dose-dependently increased CXCL17 expression by human keratinocytes in vitro. As CXCL17 mRNA expression was increased by treatment with imiquimod (IMQ), we examined the effects of CXCL17 in IMQ-induced psoriasis-like skin inflammation. Injection of recombinant CXCL17 into the ear before and during IMQ application decreased ear thickness, inflammatory cytokine expression, and the number of infiltrating cells compared with PBS injection. Flow cytometric analysis and immunofluorescent staining revealed that the numbers of MDSCs, which are CD11b
Sigma-Aldrich
Isostearic acid, ≥97% (capillary GC)
Pricing and availability is not currently available.