Merck
  • Monoaminergic modulation of GABAergic transmission onto cerebellar globular cells.
Monoaminergic modulation of GABAergic transmission onto cerebellar globular cells.
Neuropharmacology (2017-03-17)
Moritoshi Hirono, Soichi Nagao, Yuchio Yanagawa, Shiro Konishi
ABSTRACT
Cerebellar Purkinje cells (PCs) project their axon collaterals to underneath of the PC layer and make GABAergic synaptic contacts with globular cells, a subgroup of Lugaro cells. GABAergic transmission derived from the PC axon collaterals is so powerful that it could inhibit globular cells and regulate their firing patterns. However, the physiological properties and implications of the GABAergic synapses on globular cells remain unknown. Using whole-cell patch-clamp recordings from globular cells in the mouse cerebellum, we examined the monoaminergic modulation of GABAergic inputs to these cells. Application of either serotonin (5-HT) or noradrenaline (NA) excited globular cells, thereby leading to their firing. The 5-HT- and NA-induced firing was temporally confined and attenuated by GABAergic transmission, although 5-HT and NA exerted an inhibitory effect on the release of GABA from presynaptic terminals of PC axon collaterals. Agonists for 5-HT
Sigma-Aldrich
EMD386088, ≥98% (HPLC)
Supelco
Serotonin, analytical standard