Merck
  • Factors screening to statistical experimental design of racemic atenolol kinetic resolution via transesterification reaction in organic solvent using free Pseudomonas fluorescens lipase.
Factors screening to statistical experimental design of racemic atenolol kinetic resolution via transesterification reaction in organic solvent using free Pseudomonas fluorescens lipase.
Chirality (2017-04-26)
Joni Agustian, Azlina Harun Kamaruddin, Hassan Y Aboul-Enein
ABSTRACT
As the (R)-enantiomer of racemic atenolol has no β-blocking activity and no lack of side effects, switching from the racemate to the (S)-atenolol is more favorable. Transesterification of racemic atenolol using free enzymes investigated as a resource to resolve the racemate via this method is limited. Screenings of enzyme, medium, and acetyl donor were conducted first to give Pseudomonas fluorescens lipase, tetrahydrofuran, and vinyl acetate. A statistical design of the experiment was then developed using Central Composite Design on some operational factors, which resulted in the conversions of 11.70-61.91% and substrate enantiomeric excess (ee) of 7.31-100%. The quadratic models are acceptable with R
Sigma-Aldrich
(R)-(+)-Atenolol, 99%