Merck
  • Nanotribological properties of nanostructured poly(cysteine methacrylate) brushes.
Nanotribological properties of nanostructured poly(cysteine methacrylate) brushes.
Soft matter (2017-02-22)
Omed Al-Jaf, Abdullah Alswieleh, Steven P Armes, Graham J Leggett
ABSTRACT
The nanomechanical properties of zwitterionic poly(cysteine methacrylate) (PCysMA) brushes grown from planar surfaces by atom transfer radical polymerisation have been characterised by friction force microscopy (FFM). FFM provides quantitative insights into polymer structure-property relationships and in particular illuminates the dependence of brush swelling on chain packing in nanostructured materials. In ethanol, which is a poor solvent for PCysMA, a linear friction-load relationship is observed, indicating that energy dissipation occurs primarily through ploughing. In contrast, in a good solvent for PCysMA such as water, a non-linear friction-load relationship is observed that can be fitted by Derjaguin-Muller-Toporov (DMT) mechanics, suggesting that the relatively small modulus of the swollen polymer leads to a large contact area and consequently a significant shear contribution to energy dissipation. The brush grafting density was varied by using UV photolysis of C-Br bonds at 244 nm to dehalogenate the surface in a controlled fashion. The surface shear strength increases initially as the brush grafting density is reduced, but then decreases for UV doses greater than 0.5 J cm
Sigma-Aldrich
α-Bromoisobutyryl bromide, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethylphenylphosphine, 99%
Pricing and availability is not currently available.