Merck
  • Lipid droplet dynamics and insulin sensitivity upon a 5-day high-fat diet in Caucasians and South Asians.
Lipid droplet dynamics and insulin sensitivity upon a 5-day high-fat diet in Caucasians and South Asians.
Scientific reports (2017-02-15)
Anne Gemmink, Leontine E H Bakker, Bruno Guigas, Esther Kornips, Gert Schaart, A Edo Meinders, Ingrid M Jazet, Matthijs K C Hesselink
ABSTRACT
A 5-day High-Fat High-Calorie diet (HFHC-diet) reduces insulin-stimulated glucose disposal (Rd) in South Asian, but not Caucasian healthy lean males. We aimed to investigate if differences in myocellular lipid handling are underlying this differential response. A two-step hyperinsulinemic-euglycemic clamp and muscle biopsies were performed in 12 healthy lean Caucasian and South Asian males (BMI < 25 kg/m2, 19-25 years) before and after a 5-day HFHC-diet (regular diet + 375 mL cream/day; 1275 kcal/day; 94% fat). Triglyceride extractions and Western Blots for lipid droplet and mitochondrial proteins were performed. Intramyocellular lipid content and HFHC-diet response were similar between ethnicities (group effect: P = 0.094; diet effect: +~30%, P = 0.044). PLIN5 protein content increased upon the HFHC-diet (P = 0.031) and tended to be higher in South Asians (0.87 ± 0.42 AU vs. 1.35 ± 0.58 AU, P = 0.07). 4-HNE tended to increase in South Asians upon the HFHC-diet (interaction effect: P = 0.057). In Caucasians ΔPLIN5 content correlated with ΔRd (Caucasians: r = 0.756, P = 0.011; South Asians: r = -0.085, P = 0.816), while in South Asians Δ4-HNE associated with ΔPLIN5 content (Caucasians: r = 0.312, P = 0.380; South Asians: r = 0.771, P = 0.003). These data indicate that in Caucasians, PLIN5 may be protective against HFHC-diet induced insulin resistance, which for reasons not yet understood is not observed in South Asians, who possess increased lipid peroxidation levels.
Sigma-Aldrich
Monoclonal Anti-Actin (α-Sarcomeric) antibody produced in mouse, clone 5C5, ascites fluid
Sigma-Aldrich
Diethyl 3,4-pyrroledicarboxylate, 98%