Merck
  • Short-term phytotoxicity in Brassica napus (L.) in response to pre-emergently applied metazachlor: A microcosm study.
Short-term phytotoxicity in Brassica napus (L.) in response to pre-emergently applied metazachlor: A microcosm study.
Environmental toxicology and chemistry (2016-06-28)
Hanne Vercampt, Lyubka Koleva, Andon Vassilev, Jaco Vangronsveld, Ann Cuypers
ABSTRACT
In accordance with realistic application approaches, a short-term 1-factorial experiment was set up to investigate the phytotoxic impact of pre-emergent application of the chloroacetamide herbicide metazachlor on Brassica napus. In addition to morphological parameters, the underlying processes that ultimately determine the extent of herbicide-induced phytotoxicity (i.e., herbicide metabolization and cellular antioxidant defense) were examined. The present study demonstrated that metazachlor provoked fasciation of the leaves closely after emergence, which might be linked to its mode of action whereby cell division is impaired through the inhibition of very long chain fatty acid synthesis. The increased activities of antioxidative enzymes and metabolites in leaf tissue indicated the presence of reactive oxygen species under the influence of metazachlor. This resulted in oxidative damage in the form of membrane lipid peroxidation. Simultaneously, the increased activity of glutathione S-transferase and the shift in glutathione redox state suggested activation of the detoxification metabolism. This occurred, however, at the expense of growth, with a temporary reduction in plant height and weight after application. The results indicated that metazachlor disappeared within 3 mo to 4 mo after application, which resulted in the recovery of the crop. In conclusion, metazachlor induces phytotoxicity in the short term, either directly through its mode of action or indirectly through the induction of oxidative stress, which resulted in a temporary reduction in growth. Environ Toxicol Chem 2017;36:59-70. © 2016 SETAC.
Product Description
Supelco
Metazachlor, PESTANAL®, analytical standard
Pricing and availability is not currently available.