Merck
  • Electrodeposition of zinc nanoplates from an ionic liquid composed of 1-butylpyrrolidine and ZnCl
Electrodeposition of zinc nanoplates from an ionic liquid composed of 1-butylpyrrolidine and ZnCl
Dalton transactions (Cambridge, England : 2003) (2016-12-14)
Giridhar Pulletikurthi, Maryam Shapouri Ghazvini, Tong Cui, Natalia Borisenko, Timo Carstens, Andriy Borodin, Frank Endres
ABSTRACT
The mixtures of 1-butylpyrrolidine and ZnCl
Sigma-Aldrich
1-Butylpyrrolidine, 98%