Merck
  • Oxidized nucleotide insertion by pol β confounds ligation during base excision repair.
Oxidized nucleotide insertion by pol β confounds ligation during base excision repair.
Nature communications (2017-01-10)
Melike Çağlayan, Julie K Horton, Da-Peng Dai, Donna F Stefanick, Samuel H Wilson
ABSTRACT
Oxidative stress in cells can lead to accumulation of reactive oxygen species and oxidation of DNA precursors. Oxidized purine nucleotides can be inserted into DNA during replication and repair. The main pathway for correcting oxidized bases in DNA is base excision repair (BER), and in vertebrates DNA polymerase β (pol β) provides gap filling and tailoring functions. Here we report that the DNA ligation step of BER is compromised after pol β insertion of oxidized purine nucleotides into the BER intermediate in vitro. These results suggest the possibility that BER mediated toxic strand breaks are produced in cells under oxidative stress conditions. We observe enhanced cytotoxicity in oxidizing-agent treated pol β expressing mouse fibroblasts, suggesting formation of DNA strand breaks under these treatment conditions. Increased cytotoxicity following MTH1 knockout or treatment with MTH1 inhibitor suggests the oxidation of precursor nucleotides.
Sigma-Aldrich
(S)-Crizotinib, ≥98% (HPLC)
Pricing and availability is not currently available.